Learn More
Cayman Chemical BIlIrubIn asy Catlyst 15mL
SDP
Catalog No. 502942483
Encompass_Preferred
Bilirubin assay catalyst has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).
Catalog No. 50-294-2483 Supplier Cayman Chemical Supplier No. 40050615 ML
Bilirubin assay catalyst has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).