Learn More
Medchemexpress LLC Bilirubin conjugate disodium | 68683-34-1 | MFCD00044931 | 95.0% | C37H44N6Na2O10S2 | 10MG
SDP
Catalog No. 5000260151
Encompass_Preferred
Bilirubin conjugate disodium is a water-soluble, ditaurate derivative of bilirubin used as a research reagent and drug-metabolite reference for studies of heme metabolism and assay development. The material is supplied as the disodium salt (C37H44N6Na2O10S2) and is accompanied by analytical documentation for laboratory use.
  • High purity (95.0%).
  • Water-soluble disodium salt for aqueous assays.
  • Suitable as a reference standard in bilirubin and drug-metabolite studies.
  • Provided with certificate of analysis and safety data sheet.
  • Available in small quantity packages for research applications.
Catalog No. 50-002-60151 Supplier Medchemexpress LLC Supplier No. HY12983410MG
Bilirubin conjugate disodium is a water-soluble, ditaurate derivative of bilirubin used as a research reagent and drug-metabolite reference for studies of heme metabolism and assay development. The material is supplied as the disodium salt (C37H44N6Na2O10S2) and is accompanied by analytical documentation for laboratory use.
  • High purity (95.0%).
  • Water-soluble disodium salt for aqueous assays.
  • Suitable as a reference standard in bilirubin and drug-metabolite studies.
  • Provided with certificate of analysis and safety data sheet.
  • Available in small quantity packages for research applications.