Learn More
Cayman Chemical BIlIrubIn ConjugatesodIM10mg
SDP
Catalog No. 502967729
Encompass_Preferred
A stable, water-soluble ditaurate derivative of bilirubin meant to mimic endogenous bilirubin glucuronide derivatives; exhibits anti-oxidant and antimutagenic effects
Catalog No. 50-296-7729 Supplier Cayman Chemical Supplier No. 1717010 MG
Add to Cart
Add to Cart
A stable, water-soluble ditaurate derivative of bilirubin meant to mimic endogenous bilirubin glucuronide derivatives; exhibits anti-oxidant and antimutagenic effects