Learn More
Cayman Chemical Bilirubin Conjugate (sodium salt), Purity: ≥95%, CAS: 68683-34-1, 500 mg
SDP
Catalog No. 502967736
Encompass_Preferred
A stable, water-soluble ditaurate derivative of bilirubin meant to mimic endogenous bilirubin glucuronide derivatives; exhibits anti-oxidant and antimutagenic effects
Catalog No. 50-296-7736 Supplier Cayman Chemical Supplier No. 17170500 MG
A stable, water-soluble ditaurate derivative of bilirubin meant to mimic endogenous bilirubin glucuronide derivatives; exhibits anti-oxidant and antimutagenic effects