Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bilirubin, MP Biomedicals™

Catalog No. MP219947491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219947491 1 g
MP219947483 250 mg
MP219947490 500 mg
MP219947494 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219947491 Supplier MP Biomedicals, Inc Supplier No. 0219947491
Add to Cart
Add to Cart

Used to prepare standard stock solutions for color density comparison in the determination of serum bilirubin. Bilirubin, MP Biomedicals is the major end-product of heme breakdown. It is the chromophore responsible for coloration in various forms of jaund

Bilirubin is the principal pigment of bile and a constituent of many biliary calculi. It is also found in blood. The compound appears to function as an antioxidant and efficient peroxyl radical scavenger, protecting membrane lipids from oxidation by these radicals. At nanomolar concentrations it has been shown to protect neurons from oxidative damage.

  • Bilirubin is produced from Ox-gall, which is sterilized before extraction with high pressure vapor at 120°C
  • Then, the bilirubin is extracted in a continuous extraction process with chloroform as a crude product
  • Recrystallization and purification is with ethanol and chloroform
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-65-4
Molecular Formula C33H36N4O6
Molecular Weight (g/mol) 584.67
MDL Number MFCD00005499
InChI Key BPYKTIZUTYGOLE-IFADSCNNSA-N
Synonym bilirubin, hematoidin, hemetoidin, bilirubin ix-alpha, principal bile pigment, unii-rfm9x3lj49, bilirubin ixalpha, 21h-biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo, rfm9x3lj49, biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl
PubChem CID 5280352
ChEBI CHEBI:16990
IUPAC Name 3-(2-{[3-(2-carboxyethyl)-5-{[(2Z)-4-ethenyl-3-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-2-yl]methyl}-5-{[(2Z)-3-ethenyl-4-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-3-yl)propanoic acid
SMILES CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1

Specifications

Quantity 1 g
Formula Weight 584.7g/mol
Physical Form Powder
Recommended Storage Store at 0°C. Protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.