Learn More
Sigma Aldrich Fine Chemicals Biosciences Bilirubin Oxidase from Myr
Catalog No. 502902703
Encompass
We are committed to bringing younbspGreenernbspAlternative Products which adhere to one or more of The 12 Principles ofnbspGreenernbspChemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell research. for more information see the article in biofiles.Bilirubin oxidase (BOD) belongs to the sub class of multicopper oxidase family. It is usually found in fungi.
Catalog No. 50-290-2703 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B0390100UN
We are committed to bringing younbspGreenernbspAlternative Products which adhere to one or more of The 12 Principles ofnbspGreenernbspChemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell research. for more information see the article in biofiles.Bilirubin oxidase (BOD) belongs to the sub class of multicopper oxidase family. It is usually found in fungi.