Learn More
Medchemexpress LLC Bilirubin Oxidase, Myrothecium verrucaria | 80619-01-8 | 250 U
SDP
Catalog No. 5000397449
Encompass_Preferred
Bilirubin oxidase (BOD) is a multi-copper oxidase that catalyzes the oxidation of bilirubin to biliverdin, reducing molecular oxygen to water. Sourced from Myrothecium verrucaria, it plays a role in the metabolism of porphyrins and chlorophyll, making it valuable for biochemical research as an oxygen reduction catalyst.
  • Catalyzes the oxidation of bilirubin to biliverdin
  • Reduces molecular oxygen to water
  • Involved in the metabolism of porphyrins and chlorophyll
  • Acts on the CH-CH group as a donor, using oxygen as an acceptor
  • Substrates include bilirubin and O2
  • Products include biliverdin and H2O
  • Used in biochemical research
  • Appearance is a light blue to blue solid
Catalog No. 50-003-97449 Supplier Medchemexpress LLC Supplier No. 250UHYP2888
Bilirubin oxidase (BOD) is a multi-copper oxidase that catalyzes the oxidation of bilirubin to biliverdin, reducing molecular oxygen to water. Sourced from Myrothecium verrucaria, it plays a role in the metabolism of porphyrins and chlorophyll, making it valuable for biochemical research as an oxygen reduction catalyst.
  • Catalyzes the oxidation of bilirubin to biliverdin
  • Reduces molecular oxygen to water
  • Involved in the metabolism of porphyrins and chlorophyll
  • Acts on the CH-CH group as a donor, using oxygen as an acceptor
  • Substrates include bilirubin and O2
  • Products include biliverdin and H2O
  • Used in biochemical research
  • Appearance is a light blue to blue solid