Learn More
Verichem Laboratories BILIRUBIN STD KIT 10X5ML
Catalog No. NC9592200
Encompass_Preferred
Protein-based, optically clear liquid standards. Intended specifically for calibration verification. Known linear relation stated in product insert. Stable five days after opening when refrigerated. Kit includes two, five-level sets. No glycols, azides or surfactants.
Catalog No. NC9592200 Supplier Verichem Laboratories Supplier No. 9450
Protein-based, optically clear liquid standards. Intended specifically for calibration verification. Known linear relation stated in product insert. Stable five days after opening when refrigerated. Kit includes two, five-level sets. No glycols, azides or surfactants.