Learn More
Verichem Laboratories BILIRUBIN STD LVL F 2X5ML
Catalog No. NC9592201
Encompass_Preferred
This product is not sold separately and must be ordered with Bilirubin Standard Kit Direct Bilirubin values may vary with methodology and conditions of assay Identical matrix as Verichem 9450 Bilirubin Standard Kit for optimum linearity Protein based, ready-to-use liquid No glycols, azides or surfactants Concentrations verified against available NIST Standard Reference Material Stable 5 days after opening when refrigerated
Catalog No. NC9592201 Supplier Verichem Laboratories Supplier No. 9436
This product is not sold separately and must be ordered with Bilirubin Standard Kit Direct Bilirubin values may vary with methodology and conditions of assay Identical matrix as Verichem 9450 Bilirubin Standard Kit for optimum linearity Protein based, ready-to-use liquid No glycols, azides or surfactants Concentrations verified against available NIST Standard Reference Material Stable 5 days after opening when refrigerated