Learn More
Teco Diagnostics BILIRUBIN TOTAL 4X120ML ASSAYS
Catalog No. NC0714393
Encompass
For the quantitative determination of total bilirubin in human serum. 4x30mL Vials of Bilirubin, 1x30mL Vial of Bilirubin Nitrate Reagent, 1x10mL Vial of Bilirubin Calibrator
Catalog No. NC0714393 Supplier Teco Diagnostics Supplier No. B576480
For the quantitative determination of total bilirubin in human serum. 4x30mL Vials of Bilirubin, 1x30mL Vial of Bilirubin Nitrate Reagent, 1x10mL Vial of Bilirubin Calibrator