Learn More
Medchemexpress LLC Biliverdin | 114-25-0 | 70.0% | C33H34N4O6 | 10MG
SDP
Catalog No. 5000259257
Encompass_Preferred
Biliverdin is a green tetrapyrrolic bile pigment produced by heme catabolism, supplied as a research-grade reagent for biochemical and cell biology studies. It is commonly used as an endogenous metabolite in inflammation and heme metabolism research; handle protected from light and prepare DMSO stocks for in vitro work.
Catalog No. 50-002-59257 Supplier Medchemexpress LLC Supplier No. HY135005A10MG
Biliverdin is a green tetrapyrrolic bile pigment produced by heme catabolism, supplied as a research-grade reagent for biochemical and cell biology studies. It is commonly used as an endogenous metabolite in inflammation and heme metabolism research; handle protected from light and prepare DMSO stocks for in vitro work.