Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biliverdin | 114-25-0 | MFCD00870216 | 70.0% | 582.65 | C33H34N4O6 | 5 MG
SDP

Catalog No. 5000223362
Encompass_Preferred

Biliverdin is a tetrapyrrolic bile pigment produced during heme catabolism and provided as a research reagent for biochemical and cellular studies. It is investigated for antioxidant, anti-inflammatory, and immunomodulatory effects and for its role in heme metabolism and tissue protection. For research use only.

  • Endogenous metabolite used in biochemical and cell biology research.
  • Exhibits antioxidant, anti-inflammatory, and immunosuppressant properties.
  • Soluble in DMSO at about 3 mg/mL; ultrasonic warming recommended.
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.
  • Available in small milligram quantities suitable for laboratory experiments.

Catalog No. 50-002-23362 Supplier Medchemexpress LLC Supplier No. HY135005A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biliverdin is a tetrapyrrolic bile pigment produced during heme catabolism and provided as a research reagent for biochemical and cellular studies. It is investigated for antioxidant, anti-inflammatory, and immunomodulatory effects and for its role in heme metabolism and tissue protection. For research use only.

  • Endogenous metabolite used in biochemical and cell biology research.
  • Exhibits antioxidant, anti-inflammatory, and immunosuppressant properties.
  • Soluble in DMSO at about 3 mg/mL; ultrasonic warming recommended.
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.
  • Available in small milligram quantities suitable for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.