Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biliverdin Dihydrochloride, approx. 80%, MP Biomedicals™

Catalog No. ICN19488625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19488625 25 mg
ICN19488680 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19488625 Supplier MP Biomedicals, Inc Supplier No. 0219488625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55482-27-4
Molecular Formula C33H36Cl2N4O6
Molecular Weight (g/mol) 655.573
InChI Key GNLHFBAZSKVVCQ-VTURHDOZSA-N
Synonym biliverdin hydrochloride
PubChem CID 132991794
IUPAC Name 3-[(2Z)-2-[[3-(2-carboxyethyl)-5-[(E)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methylidene]-5-[(4-ethenyl-3-methyl-5-oxopyrrol-2-yl)methylidene]-4-methylpyrrol-3-yl]propanoic acid;dihydrochloride
SMILES CC1=C(C(=CC2=C(C(=C(N2)C=C3C(=C(C(=O)N3)C)C=C)C)CCC(=O)O)NC1=CC4=NC(=O)C(=C4C)C=C)CCC(=O)O.Cl.Cl

Specifications

Color Green
Assay Percent Range Approx. 80%
Quantity 25 mg
Formula Weight 655.6
Physical Form Solid
Chemical Name or Material Biliverdin Dihydrochloride
storageNote1 Protect from light.
Recommended Storage Store at +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.