Learn More
Echelon Biosciences Research Labs BILIVERDIN HYDROCHLORIDE 100mg
SDP
Supplier:  Echelon Biosciences Research Labs FH100100MG
Encompass
Biliverdin Hydrochloride is produced from the oxidation of heme in a reaction catalyzed by the enzyme heme oxygenase. Heme oxygenase has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation, reduction of inflammation, and has been associated with vascular repair. In vivo, biliverdin is reduced to bilirubin. Biliverdin Hydrochloride is soluble in basic aqueous solutions (pH > 9 for intitial dissolution) and soluble down to pH 7 once in solution as well as methanol and ethanol if made slightly basic. This item is non returnable
Catalog No. 50-720-337