Learn More
Medchemexpress LLC Biliverdin hydrochloride | 856699-18-8 | MFCD06245593 | 99.3% | C33H35ClN4O6 | 10MG
SDP
Catalog No. 5000259248
Encompass_Preferred
Biliverdin hydrochloride is the hydrochloride salt of biliverdin, a green tetrapyrrolic bile pigment formed by heme catabolism. It is provided as a water-soluble research reagent for biochemical and analytical applications, commonly used in studies of antioxidant, anti-inflammatory, and immunomodulatory activity, and as an analytical standard.
  • Water-soluble hydrochloride salt suitable for aqueous formulations.
  • Useful in biochemical assays as an antioxidant and anti-inflammatory agent.
  • Suitable as an analytical standard for LC/MS and spectroscopic methods.
  • High reported purity for reliable experimental results.
  • Available in small milligram quantities for laboratory research.
Catalog No. 50-002-59248 Supplier Medchemexpress LLC Supplier No. HY13500510MG
Biliverdin hydrochloride is the hydrochloride salt of biliverdin, a green tetrapyrrolic bile pigment formed by heme catabolism. It is provided as a water-soluble research reagent for biochemical and analytical applications, commonly used in studies of antioxidant, anti-inflammatory, and immunomodulatory activity, and as an analytical standard.
  • Water-soluble hydrochloride salt suitable for aqueous formulations.
  • Useful in biochemical assays as an antioxidant and anti-inflammatory agent.
  • Suitable as an analytical standard for LC/MS and spectroscopic methods.
  • High reported purity for reliable experimental results.
  • Available in small milligram quantities for laboratory research.