Learn More
Enzo Life Sciences Biliverdin Reductase (rat), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIOSP450D
Encompass_Preferred
Alternative name: BLVR. MW: ~33kDa. Purity: ≥95% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 1.0mM EDTA, and 5.0mM DTT. Source: Produced in E. coli. UniProt: P46844. Application: WB, Activity assay. Long Term Storage: -80°C
Catalog No. 50-667-026