Learn More
Enzo Life Sciences Biliverdin Reductase polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADIOSA450E
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant rat Biliverdin Reductase. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat E. coli. Applications: IP, WB. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7370