Learn More
Cayman Chemical BIlIverdInhydrochlorIde 100mg
SDP
Catalog No. 502984669
Encompass_Preferred
A green bile pigment produced from the oxidation of heme in a reaction catalyzed by heme oxygenase and further reduced to bilirubin by biliverdin reductase; possesses anti-mutagenic and antioxidant properties, scavenging free radicals
Catalog No. 50-298-4669 Supplier Cayman Chemical Supplier No. 19257100 MG
A green bile pigment produced from the oxidation of heme in a reaction catalyzed by heme oxygenase and further reduced to bilirubin by biliverdin reductase; possesses anti-mutagenic and antioxidant properties, scavenging free radicals