Learn More
Cayman Chemical Biliverdin (hydrochloride), Purity: ≥95%, CAS: 856699-18-8, 50 mg
SDP
Catalog No. 502984040
Encompass_Preferred
A green bile pigment produced from the oxidation of heme in a reaction catalyzed by heme oxygenase and further reduced to bilirubin by biliverdin reductase; possesses anti-mutagenic and antioxidant properties, scavenging free radicals
Catalog No. 50-298-4040 Supplier Cayman Chemical Supplier No. 1925750 MG
A green bile pigment produced from the oxidation of heme in a reaction catalyzed by heme oxygenase and further reduced to bilirubin by biliverdin reductase; possesses anti-mutagenic and antioxidant properties, scavenging free radicals