Learn More
Cayman Chemical BIlIverdIntechnIcal grade 10mg
SDP
Catalog No. 502912225
Encompass_Preferred
A bile pigment; formed via oxidative catabolism of heme by HO-1; has antioxidant activity in ORAC and TEAC assays at >0.5 µM
Catalog No. 50-291-2225 Supplier Cayman Chemical Supplier No. 3633710 MG
A bile pigment; formed via oxidative catabolism of heme by HO-1; has antioxidant activity in ORAC and TEAC assays at >0.5 µM