Learn More
Cayman Chemical BIlIvRdIn Reductase Bhumn 50ug
SDP
Catalog No. 502945508
Encompass_Preferred
Source: Active recombinant human biliverdin reductase B expressed in E. coli • Amino acids: 1-206 (full length) • MW: 22 kDa
Catalog No. 50-294-5508 Supplier Cayman Chemical Supplier No. 4047750 UG
Source: Active recombinant human biliverdin reductase B expressed in E. coli • Amino acids: 1-206 (full length) • MW: 22 kDa