Learn More
Chemglass Life Sciences Bill-Board Complete Set
SDP
Catalog No. 5000980708
Encompass_Preferred
The Bill-Board set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i. e. simultaneous silica gel chromatography). complete Set Consists of: 6 each: CG-1869-08 Glass Vessels 1 each: CG-1869-10 6 Position Top Plate 1 each: CG-1869-12 Drain Tray 1 each: CG-1869-14 Vial Collection Rack 6 each: CG-1869-50 4 Dram, 25 x 52mm Collection Vials. Bill-Board Individual Components: CG-1869-08: Glass Reaction Vessels: Vessels are 75mm long with 13-425 Threads on either end. A fr
Catalog No. 50-009-80708 Supplier Chemglass Life Sciences Supplier No. CG186901
The Bill-Board set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i. e. simultaneous silica gel chromatography). complete Set Consists of: 6 each: CG-1869-08 Glass Vessels 1 each: CG-1869-10 6 Position Top Plate 1 each: CG-1869-12 Drain Tray 1 each: CG-1869-14 Vial Collection Rack 6 each: CG-1869-50 4 Dram, 25 x 52mm Collection Vials. Bill-Board Individual Components: CG-1869-08: Glass Reaction Vessels: Vessels are 75mm long with 13-425 Threads on either end. A fr