Learn More
AdipoGen Bilobalide (50 mg)
SDP
Supplier:  AdipoGen AGCN20026M050
Encompass_Preferred
Chemical. CAS: 33570-04-6. Formula: C15H18O8. MW: 326.3. Isolated from Ginko biloba. Neuroprotective. Mitochondrial gene expression regulator. ROS scavenger. Competitive GABA(A) receptor antagonist. Apoptosis inhibitor. CREB phosphorylation enhancer. Stimulates neurogenesis and synaptogenesis. Hepatic cytochrome P450 inducer. Activates the phosphatidylinositol 3-kinase (PI3K) dependent pathway. Potent anti-inflammatory and antihyperalgesic agent.
Catalog No. 50-205-3372