Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Bilobalide, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013957 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3957 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3957 Supplier MilliporeSigma Supelco Supplier No. 7959310MG
Add to Cart
Add to Cart

(−)-Bilobalide is a Ginkgo terpenoid, available as the main pharmacological component in Ginkgo biloba extract. It exhibits neuroprotectant activity.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 33570-04-6
Physical Form Neat
Linear Formula C15H18O8
MDL Number MFCD00132880
Synonym Bilobalide
Type Analytical Standard
Quantity 10 mg
Formula Weight 326.30
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.