Learn More
ATCC Bilophila wadsworthia
Catalog No. 502384705
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Bilophila wadsworthia strain WAL 7959 [Lab 88-130H] is a whole-genome sequenced bacterial type strain that was isolated from a perforated appendiceal abscess. This organism is a strict anaerobe.
Catalog No. 50-238-4705 Supplier ATCC Supplier No. 49260
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Bilophila wadsworthia strain WAL 7959 [Lab 88-130H] is a whole-genome sequenced bacterial type strain that was isolated from a perforated appendiceal abscess. This organism is a strict anaerobe.