Learn More
Medchemexpress LLC BIM 23042 trifluoroacetate | 00-00-0 | 96.2% | 1192.45 g/mol (free base) | C63H73N11O9S2.xC2HF3O2 | 5 MG
SDP
Catalog No. 5000244877
Encompass_Preferred
BIM 23042 TFA is a somatostatin-like octapeptide analogue that functions as a selective antagonist of the neuromedin B receptor (NMB-R/BB1). Supplied as the trifluoroacetate (TFA) salt at high purity, it is intended for research use in receptor pharmacology and related biochemical assays.
  • Selective antagonist of neuromedin B receptor (BB1).
  • TFA salt form for improved handling and stability.
  • High purity suitable for receptor pharmacology and calcium mobilization assays.
  • Supplied as a small, lyophilized quantity for laboratory research use.
  • Molecular weight and elemental composition available in the product documentation.
Catalog No. 50-002-44877 Supplier Medchemexpress LLC Supplier No. HY103277A5MG
BIM 23042 TFA is a somatostatin-like octapeptide analogue that functions as a selective antagonist of the neuromedin B receptor (NMB-R/BB1). Supplied as the trifluoroacetate (TFA) salt at high purity, it is intended for research use in receptor pharmacology and related biochemical assays.
  • Selective antagonist of neuromedin B receptor (BB1).
  • TFA salt form for improved handling and stability.
  • High purity suitable for receptor pharmacology and calcium mobilization assays.
  • Supplied as a small, lyophilized quantity for laboratory research use.
  • Molecular weight and elemental composition available in the product documentation.