Learn More
Medchemexpress LLC BIM 23042 (trifluoroacetate) | 00-00-0 | MFCD32666112 | 96.2% | 1192.45 g/mol | C63H73N11O9S2·xC2HF3O2 | 1 MG
SDP
Catalog No. 5000247560
Encompass_Preferred
BIM 23042 (trifluoroacetate) is a somatostatin-like octapeptide analogue that acts as a selective neuromedin B receptor (NMB-R/BB1) antagonist. It is supplied as the trifluoroacetate salt in solid form for research use in receptor pharmacology and intracellular calcium signaling studies.
  • Selective neuromedin B receptor (NMB-R/BB1) antagonist.
  • Somatostatin-like octapeptide structure as the trifluoroacetate salt.
  • Purity approximately 96.2% by supplier analysis.
  • White to off-white solid powder suitable for laboratory use.
  • Store sealed away from moisture and light; powder stable at -80°C (2 years) or -20°C (1 year).
Catalog No. 50-002-47560 Supplier Medchemexpress LLC Supplier No. HY103277A1MG
BIM 23042 (trifluoroacetate) is a somatostatin-like octapeptide analogue that acts as a selective neuromedin B receptor (NMB-R/BB1) antagonist. It is supplied as the trifluoroacetate salt in solid form for research use in receptor pharmacology and intracellular calcium signaling studies.
  • Selective neuromedin B receptor (NMB-R/BB1) antagonist.
  • Somatostatin-like octapeptide structure as the trifluoroacetate salt.
  • Purity approximately 96.2% by supplier analysis.
  • White to off-white solid powder suitable for laboratory use.
  • Store sealed away from moisture and light; powder stable at -80°C (2 years) or -20°C (1 year).