Learn More
Medchemexpress LLC BIM-23190 hydrochloride | 182153-96-4 | 99.33% | 1238.91 | 10 MG
SDP
Catalog No. 5000398667
Encompass_Preferred
BIM-23190 hydrochloride is a somatostatin analog and a selective SSTR2 and SSTR5 agonist, with Ki values of 0.34 nM for SSTR2 and 11.1 nM for SSTR5. It can be used in studies for cancer and acromegaly.
  • Somatostatin analog
  • Selective SSTR2 and SSTR5 agonist
  • Exhibits anti-tumor activity
  • Can be used in the study of cancer and acromegaly
Catalog No. 50-003-98667 Supplier Medchemexpress LLC Supplier No. HYP3124A10MG
BIM-23190 hydrochloride is a somatostatin analog and a selective SSTR2 and SSTR5 agonist, with Ki values of 0.34 nM for SSTR2 and 11.1 nM for SSTR5. It can be used in studies for cancer and acromegaly.
  • Somatostatin analog
  • Selective SSTR2 and SSTR5 agonist
  • Exhibits anti-tumor activity
  • Can be used in the study of cancer and acromegaly