Learn More
Medchemexpress LLC HY-P3124A 5mg Medchemexpress, BIM-23190 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP3124A5MG
Encompass_Preferred
Medchemexpress, HY-P3124A 5mg BIM-23190 (hydrochloride) CAS: BIM-23190 hydrochloride, a somatostatin analog, a selective SSRT2 and SSRT5 agonist, exhibits Ki values of 0.34 nM and 11.1 nM for SSTR2 and SSTR5, respectively. BIM-23190 can be used in the study for cancer and acromegaly. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3094