Learn More
TARGETMOL CHEMICALS INC BIM-46187 4HCL 5MG
SDP
Catalog No. 502581170
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIM-46187 4HCl (PH-064 4HCl) is a heterotrimeric G-protein complex inhibitor. purity: 99%
Catalog No. 50-258-1170 Supplier TARGETMOL CHEMICALS INC Supplier No. T16519L5MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIM-46187 4HCl (PH-064 4HCl) is a heterotrimeric G-protein complex inhibitor. purity: 99%