Learn More
Medchemexpress LLC Bim BH3 | 505070-06-4 | 99.7% | 2444.77 | 5 MG
SDP
Catalog No. 5000422340
Encompass_Preferred
Bim BH3 is a biologically active peptide that belongs to the pro-apoptotic Bcl-2 family of proteins. It is for research use only and not sold to patients.
  • Biologically active peptide
  • Belongs to the pro-apoptotic Bcl-2 family of proteins
  • Intended for research use only
  • Functions as an inhibitor
Catalog No. 50-004-22340 Supplier Medchemexpress LLC Supplier No. HYP15275MG
Bim BH3 is a biologically active peptide that belongs to the pro-apoptotic Bcl-2 family of proteins. It is for research use only and not sold to patients.
  • Biologically active peptide
  • Belongs to the pro-apoptotic Bcl-2 family of proteins
  • Intended for research use only
  • Functions as an inhibitor