Learn More
Enzo Life Sciences Bim BOD polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP330E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the middle of human Bim/BOD. UniProt ID: O43521. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (20µg/ml) Western Blot (1µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7161