Learn More
AdipoGen Bisindolylmaleimide IX . methanesulfonate (5 mg)
SDP
Supplier:  AdipoGen AGCR10111M005
Encompass_Preferred
Chemical. CAS: 138489-18-6. Formula: C25H23N5O2S . CH4O3S. MW: 457.6 . 96.1. Selective and cell permeable protein kinase C (PKC) inhibitor. Inhibits the stimulation of insulin secretion by glucose. Inhibits T cell activation. Apoptosis inducer. Potent glycogen synthase kinase-3 (GSK-3) inhibitor. Transcription inhibitor. Induces TNF receptor family-mediated cell death. Pim-1 kinase inhibitor. Antitumor compound.
Catalog No. 50-205-3837