Learn More
AdipoGen Bisindolylmaleimide X . hydrochloride (1 mg)
SDP
Supplier:  AdipoGen AGCR10113M001
Encompass_Preferred
Chemical. CAS: 145317-11-9131848-97-0 (free base). Formula: C26H24N4O2 . HCl. MW: 424.5 . 36.5. Selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2). Inhibits superoxide generation in human neutrophils.
Catalog No. 50-205-3840