Learn More
AdipoGen Bisindolylmaleimide XI . hydrochloride (1 mg)
SDP
Supplier:  AdipoGen AGCR10109M001
Encompass_Preferred
Chemical. CAS: 145333-02-4. Formula: C28H28N4O2 . HCl. MW: 452.5 . 36.5. Selective cell permeable protein kinase C (PKC) inhibitor. G protein-coupled receptor kinase (GRK-5) inhibitor. Less potent GRK-2 or GRK-3 inhibitor. Prevents T cell-driven chronic inflammatory responses in vivo.
Catalog No. 50-205-3835