Learn More
Enzo Life Sciences Bimatoprost (10 mg). CAS: 155206-00-1
SDP
Supplier:  Enzo Life Sciences BMLRA1210010
Encompass_Preferred
Stable analog of PGF2α. Agonist at FP receptors (after in vivo hydrolysis to the free acid). It is an efficacious treatment for glaucoma. It also displays an unusual eye lash lengthening effect. Alternative name: (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]-N-ethylhept-5-enamide. Purity: ≥97%. Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1730