Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bimatoprost | 155206-00-1 | 99.9% | 5 MG
SDP

Catalog No. 502031031
Encompass_Preferred

Bimatoprost, a prostaglandin analog, is used in the study of open-angle glaucoma, ocular hypertension, and other forms of glaucoma. It reduces intraocular pressure by regulating scleral and trabecular outflow. Topical application can induce fat atrophy and cause a deepening of the eyelid sulcus. This product appears as a white to off-white solid with a purity of 99.94%.

  • Reduces intraocular pressure
  • Used for open-angle glaucoma and ocular hypertension studies
  • Induces fat atrophy
  • Purity: 99.94%
  • Soluble in Methanol and DMSO
  • Molecular weight: 415.57
  • Chemical formula: C25H37NO4
  • Powder storage: -20°C for 3 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-203-1031 Supplier Medchemexpress LLC Supplier No. HYB01915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bimatoprost, a prostaglandin analog, is used in the study of open-angle glaucoma, ocular hypertension, and other forms of glaucoma. It reduces intraocular pressure by regulating scleral and trabecular outflow. Topical application can induce fat atrophy and cause a deepening of the eyelid sulcus. This product appears as a white to off-white solid with a purity of 99.94%.

  • Reduces intraocular pressure
  • Used for open-angle glaucoma and ocular hypertension studies
  • Induces fat atrophy
  • Purity: 99.94%
  • Soluble in Methanol and DMSO
  • Molecular weight: 415.57
  • Chemical formula: C25H37NO4
  • Powder storage: -20°C for 3 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.