Learn More
TARGETMOL CHEMICALS INC Bimatoprost acid 1MG
SDP
Catalog No. 502877450
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bimatoprost acid (17-phenyl trinor PGF2alpha) is a metabolically stable analog of PGF2alpha and has a potential antagonistic activity for the FP receptor. It has a relative potency of 756% compared to PGF2alpha for binding to the FP receptor on ovine luteal cells. Purity 99.29%
Catalog No. 50-287-7450 Supplier TARGETMOL CHEMICALS INC Supplier No. T209391MG
Add to Cart
Add to Cart
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bimatoprost acid (17-phenyl trinor PGF2alpha) is a metabolically stable analog of PGF2alpha and has a potential antagonistic activity for the FP receptor. It has a relative potency of 756% compared to PGF2alpha for binding to the FP receptor on ovine luteal cells. Purity 99.29%