Learn More
Medchemexpress LLC Bimatoprost-d5 | 155206-00-1 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0191S5MG
Encompass_Preferred
Bimatoprost-d5 is a deuterium labeled Bimatoprost, a prostaglandin analog and a topical hypotensive agent frequently used for treating ocular hypertension and glaucoma. It also has an antiadipogenic effect.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05759