Learn More
Medchemexpress LLC Bimatoprost-d5 (AGN 192024 d5) | 155206-00-1 | 98.2% | 1 MG
SDP
Catalog No. 5000407417
Encompass_Preferred
Bimatoprost-d5 is a deuterium labeled Bimatoprost, functioning as a prostaglandin analog and topical hypotensive agent. It is primarily used for treating ocular hypertension and glaucoma, and also exhibits an antiadipogenic effect. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Used as a prostaglandin analog and topical hypotensive agent
  • Effective in treating ocular hypertension and glaucoma
  • Exhibits antiadipogenic properties
  • Suitable for research applications as a tracer or internal standard
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-004-07417 Supplier Medchemexpress LLC Supplier No. HYB0191S1MG
Bimatoprost-d5 is a deuterium labeled Bimatoprost, functioning as a prostaglandin analog and topical hypotensive agent. It is primarily used for treating ocular hypertension and glaucoma, and also exhibits an antiadipogenic effect. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Used as a prostaglandin analog and topical hypotensive agent
  • Effective in treating ocular hypertension and glaucoma
  • Exhibits antiadipogenic properties
  • Suitable for research applications as a tracer or internal standard
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month