Learn More
Selleck Chemical LLC Bimatoprost 50mg 155206-00-1
SDP
Catalog No. 507517
Encompass_Preferred
Bimatoprost is a prostaglandin analog used topically (as eye drops) to control the progression of glaucoma and in the management of ocular hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-751-7 Supplier Selleck Chemical LLC Supplier No. S140750MG
Bimatoprost is a prostaglandin analog used topically (as eye drops) to control the progression of glaucoma and in the management of ocular hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.