Learn More
Medchemexpress LLC BIMAX1 (acetate) | 96.1% | 3542.03 (free base) | 10 MG
SDP
Catalog No. 5000423691
Encompass_Preferred
BIMAX1 acetate is a tool peptide used to investigate the interaction of RBBP4 with importin β1 and its effects on the nuclear import pathway. It can be employed to study RBBP4's role in regulating nuclear import efficiency and cellular senescence.
  • Utilized for investigating RBBP4 and importin β1 interactions
  • Helps study nuclear import efficiency
  • Can be used to research cellular senescence
  • Appears as a white to off-white solid
Catalog No. 50-004-23691 Supplier Medchemexpress LLC Supplier No. HYP10598A10MG
BIMAX1 acetate is a tool peptide used to investigate the interaction of RBBP4 with importin β1 and its effects on the nuclear import pathway. It can be employed to study RBBP4's role in regulating nuclear import efficiency and cellular senescence.
  • Utilized for investigating RBBP4 and importin β1 interactions
  • Helps study nuclear import efficiency
  • Can be used to research cellular senescence
  • Appears as a white to off-white solid