Learn More
Medchemexpress LLC Bimax2 acetate | 98.7% | 3561.98 (free base) | 10 MG
SDP
Catalog No. 5000425075
Encompass_Preferred
Bimax2 acetate is a high affinity nuclear localization signal (NLS) peptide. It mimics the activity of the classical nuclear localization signal (cNLS) and competitively binds to importin α, thereby inhibiting the binding of cNLS-cargo proteins to importin α. It can be used to study the role of RBBP4 in regulating nuclear import efficiency and cell senescence.
  • High affinity nuclear localization signal (NLS) peptide
  • Mimics classical nuclear localization signal (cNLS) activity
  • Competitively binds to importin α
  • Inhibits binding of cNLS-cargo proteins to importin α
  • Useful for studying the role of RBBP4 in regulating nuclear import efficiency and cell senescence
  • White to off-white solid
  • Soluble in H2O at ≥ 100 mg/mL
  • For research use only
Catalog No. 50-004-25075 Supplier Medchemexpress LLC Supplier No. HYP10600A10MG
Bimax2 acetate is a high affinity nuclear localization signal (NLS) peptide. It mimics the activity of the classical nuclear localization signal (cNLS) and competitively binds to importin α, thereby inhibiting the binding of cNLS-cargo proteins to importin α. It can be used to study the role of RBBP4 in regulating nuclear import efficiency and cell senescence.
  • High affinity nuclear localization signal (NLS) peptide
  • Mimics classical nuclear localization signal (cNLS) activity
  • Competitively binds to importin α
  • Inhibits binding of cNLS-cargo proteins to importin α
  • Useful for studying the role of RBBP4 in regulating nuclear import efficiency and cell senescence
  • White to off-white solid
  • Soluble in H2O at ≥ 100 mg/mL
  • For research use only