Learn More
Cole-Parmer Digi-Sense Soil Thermometer, 48" L x 0.25" Dia; 0-200F/ -15-90C
Catalog No. 501961599 Shop All Cole-Parmer Products
Encompass_Preferred
Thermometer is ideal for measuring core temperatures in compost piles, hay, soil, or grains. 3" diameter dial for easy reading. Calibrate with external reset screw. Thermometers meet ASME B40.3 standards for leak- and fog-proof operation.Tips are 0.25" dia for fast response.
Catalog No. 50-196-1599 Supplier Cole-Parmer Supplier No. 9058730
Thermometer is ideal for measuring core temperatures in compost piles, hay, soil, or grains. 3" diameter dial for easy reading. Calibrate with external reset screw. Thermometers meet ASME B40.3 standards for leak- and fog-proof operation.Tips are 0.25" dia for fast response.