Learn More
Cayman Chemical BImIralIsIb 10mg
SDP
Catalog No. 502986965
Encompass_Preferred
A pan inhibitor of PI3Ks and an inhibitor of mTOR (IC50s = 33, 661, 708, 451, and 89 nM for PI3Kα, PI3Kβ, PI3Kγ, PI3Kδ, and mTOR, respectively); selective for these kinases over a panel of cell surface and nuclear receptors, membrane channels, transporters, kinases, proteases, and phosphodiesterases at a concentration of 10 μM; has anticancer activity (average GI50 = 0.7 μM across the NCI 60 human cancer cell line panel); reduces tumor growth in a dose-dependent manner in a PC3 prostate cancer mouse xenograft model
Catalog No. 50-298-6965 Supplier Cayman Chemical Supplier No. 2344110 MG
A pan inhibitor of PI3Ks and an inhibitor of mTOR (IC50s = 33, 661, 708, 451, and 89 nM for PI3Kα, PI3Kβ, PI3Kγ, PI3Kδ, and mTOR, respectively); selective for these kinases over a panel of cell surface and nuclear receptors, membrane channels, transporters, kinases, proteases, and phosphodiesterases at a concentration of 10 μM; has anticancer activity (average GI50 = 0.7 μM across the NCI 60 human cancer cell line panel); reduces tumor growth in a dose-dependent manner in a PC3 prostate cancer mouse xenograft model