Learn More
Cayman Chemical BimS BH3 (51-76) (human) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502914825
Encompass_Preferred
A Bim-derived peptide; binds to Mcl-1, Bcl-xL, Bcl-2, and Bcl-W (IC50s = 18, 10, 7, and 18 nM, respectively); induces cytochrome c release from mitochondria derived from bcl-x-/- MEFs
Catalog No. 50-291-4825 Supplier Cayman Chemical Supplier No. 3696310 MG
A Bim-derived peptide; binds to Mcl-1, Bcl-xL, Bcl-2, and Bcl-W (IC50s = 18, 10, 7, and 18 nM, respectively); induces cytochrome c release from mitochondria derived from bcl-x-/- MEFs