Learn More
Enzo Life Sciences BimS/EL/L monoclonal antibody (10B12) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804528C100
Encompass_Preferred
Clone: 10B12. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 20-40 (Q20PAERPPQLRPGAPTSLQTEP40) of mouse BimL. Species reactivity: Human, Mouse, Rat, Monkey, Dog. Specificity: Recognizes an epitope within aa 20-40 of BimL, BimEL, and BimS. Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified. Applications: ELISA, Flow Cytometry, ICC, IHC, IP, WB. Formulation: Liquid. Protein G affinity purified antibody in PBS containing 25% glycerol and 0.02% sodium azide.
Catalog No. 50-200-9936