Learn More
Enzo Life Sciences BimS/EL/L monoclonal antibody (14A8) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804351C100
Encompass_Preferred
Clone: 14A8. Isotype: IgG2a. Immunogen: Recombinant mouse BimS. Species reactivity: Human, Mouse Monkey. Specificity: Recognizes BimL, BimEL and BimS. Applications: Flow Cytometry, ICC, IHC (PS), IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-200-9877