Learn More
AdipoGen BimS EL L mAb 3C5
SDP
Supplier:  AdipoGen AG20T0142C100
Encompass_Preferred
Monoclonal Antibody. Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL ~19kDa, BimEL ~23kDa, and BimS ~16kDa. Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region. Isotype Rat IgG2akappa. Clone 3C5. Applications ELISA, FACS, ICC, IHC, IP, WB. Liquid. In PBS containing 0.02% sodium azide and 25% glycerol. Bim belongs to the Bcl-2 protein family. Bcl-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a Bcl-2 homology domain 3 BH3. It has been shown to interact with other members of the Bcl-2 protein family BCL2, BCL2L1/BCL-XL and MCL1 and to act as an apoptotic activator, inducing apoptosis and anoikis. Several variants have been identified.
Catalog No. 50-256-3338