Learn More
Enzo Life Sciences BimS/EL/L monoclonal antibody (3C5) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804527C100
Encompass_Preferred
Clone: 3C5. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 20-40 (Q20PAERPPQLRPGAPTSLQTEP40) of mouse BimL. Species reactivity: Human, Mouse, Rat, Monkey, Dog. Specificity: Recognizes an epitope within aa 20-40 of imL (~19kDa), BimEL (~23kDa), and BimS (~16kDa). Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified. Applications: ELISA, Flow Cytometry, ICC, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide and 25% glycerol.
Catalog No. 50-200-9935